missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Carbamazepin | |
| 191°C | |
| C15H12N2O | |
| 10 g | |
| FFGPTBGBLSHEPO-UHFFFAOYSA-N | |
| 2-azatricyclo[9.4.0.0³,â¸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide | |
| 2554 | |
| 236.27 | |
| Crystalline Powder |
| 298-46-4 | |
| White | |
| MFCD00005073 | |
| carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston | |
| NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12 | |
| 236.27 | |
| CHEBI:3387 | |
| ≥97.0% (N) |
Chemical Identifiers
| 298-46-4 | |
| 236.27 | |
| FFGPTBGBLSHEPO-UHFFFAOYSA-N | |
| 2554 | |
| 2-azatricyclo[9.4.0.0³,â¸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide |
| C15H12N2O | |
| MFCD00005073 | |
| carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston | |
| CHEBI:3387 | |
| NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12 |
Safety and Handling
RTECSNumber : HN8225000
TSCA : No