missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepin 97.0+%, TCI America™

Catalog No. C109525G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C109525G 25 g
C109510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C109525G Supplier TCI America Supplier No. C109525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 298-46-4
Molecular Formula C15H12N2O
Molecular Weight (g/mol) 236.27
MDL Number MFCD00005073
InChI Key FFGPTBGBLSHEPO-UHFFFAOYSA-N
Synonym carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston
PubChem CID 2554
ChEBI CHEBI:3387
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
SMILES NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Melting Point 191°C
Color White
Quantity 25 g
Formula Weight 236.27
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Carbamazepin
RTECSNumber HN8225000
TSCA No