missing translation for 'onlineSavingsMsg'
Learn More

Carbamate Pesticides - WS, MilliporeSigma™ Supelco™

Catalog No. 111017319
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017319 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017319 Supplier Merck Emd Millipore Supplier No. PE15072ML

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Featured Industries: Environmental

Specifications

CAS 67-56-1
Flash Point 11°C - closed cup - Solvent
Type Proficiency Testing Material
Quantity 2 mL