missing translation for 'onlineSavingsMsg'
Learn More

Carbadox, 98+%

Catalog No. AAJ6028414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6028414 25 g
AAJ6028422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6028414 Supplier Thermo Scientific Chemicals Supplier No. J6028414

CAS: 6804-07-5 | C13H10BrNO | 276.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

Solubility
Soluble in 1N NaOH (50 mg/ml), and water (partly).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 6804-07-5
MDL Number MFCD00057293

Specifications

Melting Point 239°C to 240°C
Color Yellow
Flash Point 18°C (64°F)
Molecular Formula C13H10BrNO
Quantity 25 g
UN Number UN1325
Merck Index 14,1780
Synonym 4-bromobenzanilide, n-phenyl 4-bromobenzamide, benzamide, 4-bromo-n-phenyl, 4-bromo-n-phenyl-benzamide, 4-bromo-n-phenyl-menzamide, maybridge1_004278, n1-phenyl-4-bromobenzamide, 4-bromophenyl-n-benzamide
Solubility Information Soluble in 1N NaOH (50mg/ml),and water (partly).
InChI Key QVTGXXQAVWWJRO-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Br
IUPAC Name 4-bromo-N-phenylbenzamide
Molecular Weight (g/mol) 276.13
PubChem CID 690966
Formula Weight 262.22
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Carbadox
Show More Show Less
Hazard Category H228-H302-H350
Hazard Statement GHS H Statement
H228-H350-H302
Flammable solid.
May cause cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P210-P240-P241-P264b-P270-P281-P301+P312-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
RTECSNumber FE2779000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only