missing translation for 'onlineSavingsMsg'
Learn More

Carbacyclin, 99%, Thermo Scientific Chemicals

Catalog No. p-7057709
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67199EXD 0.5 mg
AAJ67199MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67199EXD Supplier Thermo Scientific Chemicals Supplier No. J67199EXD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A stable prostacyclin analog found to inhibit platelet aggregation induced by collagen or ADP

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69552-46-1
Molecular Formula C21H34O4
Molecular Weight (g/mol) 350.50
MDL Number MFCD00210824
InChI Key XZFRIPGNUQRGPI-WBQKLGIQSA-N
Synonym carboprostacyclin, carbacyclin, carbaprostacyclin, carbacycline, carba pgx, 6a-carba-pgi2, 6,9-methano pgi2, 6,9-methano-pgi2, 6,9-methanoprostaglandin i2, 5e-6a-carba-prostaglandin i2
PubChem CID 133126726
IUPAC Name 5-[(2E,3aS,4R,5R,6aS)-5-hydroxy-4-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-octahydropentalen-2-ylidene]pentanoic acid
SMILES CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)C[C@@H]2C\C(C[C@H]12)=C/CCCC(O)=O

Specifications

Melting Point 65°C to 67°C
Color White
Quantity 0.5 mg
Solubility Information Soluble at 20mg/ml in ethanol.
Formula Weight 350.5
Percent Purity 99%
Physical Form Solid
Chemical Name or Material Carbacyclin
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only