accessibility menu, dialog, popup

UserName

CAS RN 69552-46-1

IUPAC Name: 5-[(2E,3aS,4R,5R,6aS)-5-hydroxy-4-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-octahydropentalen-2-ylidene]pentanoic acid
Synonyms: 5-[(2E,3aS,4R,5R,6aS)-5-hydroxy-4-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-hexahydro-1H-pentalen-2-ylidene]pentanoic acid
Molecular Weight (g/mol): 350.5
Molecular Formula: C21H34O4
InChi Key: XZFRIPGNUQRGPI-WBQKLGIQSA-N
SMILES: CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)C[C@@H]2C\C(C[C@H]12)=C/CCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results