missing translation for 'onlineSavingsMsg'
Learn More

Carbachol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019243
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019243 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019243 Supplier Merck Emd Millipore Supplier No. PHR1511500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Carbachol, a synthetic derivative of choline, is a highly efficient miotic agent, which can be used for a long-term therapy of non-congestive wide-angle glaucoma. Its mode of action is primarily stimulating muscarinic receptors and releasing acetylcholine at certain neuroeffector junctions and ganglia. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 51-83-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula NH2 COOCH2 CH2 N(Cl)(CH3)3
MDL Number MFCD00012011
Synonym Carbamylcholine chloride; Carbachol
Type Certified Reference Material
Quantity 500 mg
Formula Weight 182.65
Grade Certified Reference Material