missing translation for 'onlineSavingsMsg'
Learn More

Carazolol, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014724
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014724 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014724 Supplier Merck Emd Millipore Supplier No. 5378710MG
Only null left

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706030, T706025

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 57775-29-8
Physical Form Neat
Linear Formula C18H22N2O2
MDL Number MFCD00864588
Melting Point 133°C to 137°C
Synonym 1-(Carbazol-4-yloxy)-3-(isopropylamino)-2-propanol; 4-(2-Hydroxy-3-isopropylamino-propoxy)carbazole
Type Analytical Standard
Quantity 10 mg
Formula Weight 298.38
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Show More Show Less