Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG4 (Human) Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG4 (Hu) Conjugate consists of a 12 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG4 subclass.
$1,557.23 - $5,484.56
Specifications
| Product Type | Biotin Anti-IgG4 Conjugate |
|---|---|
| Conjugate | Biotin and Other Haptens |
| Product Line | CaptureSelect |
| Content And Storage | Store in freezer (-5°C to -30°C). |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
7102902100
![]() |
Thermo Scientific™
7102902100 |
100 μg |
Each for $1,557.23
|
|
|||||
7102902500
![]() |
Thermo Scientific™
7102902500 |
500 μg |
Each for $5,484.56
|
|
|||||
Description
CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate consists of a 12 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG4 subclass. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate allows you to:
- Detect, quantitate, and characterize human IgG4 antibodies
- Avoid cross-binding with other IgG subclasses like IgG1, 2 and 3 and isotypes like IgM and IgA including IgG from bovine sources such as FCS
Applications for CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
| Biotin Anti-IgG4 Conjugate | |
| Biotin and Other Haptens | |
| CaptureSelect | |
| Store in freezer (-5°C to -30°C). |