Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-AAV8 Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-AAV8 Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity to adeno-associated virus particles of serotype 8 (AAV8).
$2058.15 - $7256.63
Specifications
| Content And Storage | Store in freezer (-5°C to -30°C). |
|---|---|
| Product Line | CaptureSelect |
| Ligand Type | Antibody |
| Shipping Condition | Dry Ice |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
71-033-82100
|
Thermo Scientific™
7103382100 |
100 μg |
Each for $2,058.15
|
|
|||||
|
71-033-82500
|
Thermo Scientific™
7103382500 |
500 μg |
Each for $7,256.63
|
|
|||||
Description
CaptureSelect Biotin Anti-AAV8 Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity to adeno-associated virus particles of serotype 8 (AAV8). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize AAV8 particles.
Applications for CaptureSelect Biotin Anti-AAV8 Conjugate include ELISA, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
| Store in freezer (-5°C to -30°C). | |
| CaptureSelect | |
| Antibody | |
| Dry Ice |