Learn More
Thermo Scientific™ CaptureSelect™ AAVX Ligand Leakage ELISA Kit

Description
CaptureSelect AAVX Ligand Leakage ELISA Kit is a ready-to-use kit specifically designed to detect possible leached ligand present in adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Resin. Although the ligand is covalently coupled, it is possible for the ligand to leach and co-elute with the purified AAV. This fully assembled kit consists of a pre-coated anti-AAVX ligand strip plate, buffers, and optimal reaction solutions, enabling a robust ELISA assay to measure potential ligand leakage.
Key features:
- Reduced assay operating time, due to the pre-coating and pre-blocking of the strip plate
- Quick and easy-to-set-up assay procedure
- Sensitive and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL
The CaptureSelect AAVX Ligand Leakage ELISA Kit can be used as a tool to aid in optimal purification process development and routine quality control of in-process streams as well as final product. The kit is manufactured in an ISO-certified manufacturing site and is ISO 9001-certified.
Specifications
Specifications
| Content And Storage | • Pre-coated anti-AAVX ligand strip plate • Wash/dilutionbuffer (10X) • AAV standard/coating and biotin reagents • Streptavidin-HRP conjugate reagent • TMB substrate • Stop solution Store at 2–8°C. |
| Assay | 96-well |
| Detection Method | ELISA |
| Product Line | CaptureSelect |
| Product Type | ELISA Kit |
| For Use With (Application) | Detection of possible leached ligand |
| Quantity | 1 kit |
| Shipping Condition | 2°C to 8°C |
| Target | Anti AAVX ligand |
| Format | 96-well plate: 8-well strip plate, assembled 12 strips per plate |