missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Captopril | |
| 106°C | |
| C9H15NO3S | |
| 5 g | |
| FAKRSMQSSFJEIM-RQJHMYQMSA-N | |
| (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid | |
| 44093 | |
| 217.28 | |
| Crystalline Powder |
| 62571-86-2 | |
| White | |
| MFCD00168073 | |
| captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril | |
| CC(CS)C(=O)N1CCCC1C(=O)O | |
| 217.283 | |
| CHEBI:3380 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 62571-86-2 | |
| 217.283 | |
| FAKRSMQSSFJEIM-RQJHMYQMSA-N | |
| 44093 | |
| (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid |
| C9H15NO3S | |
| MFCD00168073 | |
| captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril | |
| CHEBI:3380 | |
| CC(CS)C(=O)N1CCCC1C(=O)O |
Safety and Handling
RTECSNumber : UY0550000
TSCA : No