missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Captopril, 98%

Catalog No. AC343700050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC343700050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC343700050 Supplier Thermo Scientific Chemicals Supplier No. 343700050
Only null left

CAS: 62571-86-2 | C9H15NO3S | 217.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62571-86-2
Molecular Formula C9H15NO3S
Molecular Weight (g/mol) 217.28
MDL Number MFCD00168073
InChI Key FAKRSMQSSFJEIM-RQJHMYQMSA-N
Synonym captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril
PubChem CID 44093
ChEBI CHEBI:3380
IUPAC Name (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
SMILES CC(CS)C(=O)N1CCCC1C(=O)O

Specifications

Melting Point 103.0°C to 108.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 1776
Specific Rotation − 129.50
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohol,chloroform,methylenech,loride
Formula Weight 217.28
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Captopril, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes serious eye damage.
May damage the unborn child.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
EINECSNumber 263-607-1
RTECSNumber UY0550000

RUO – Research Use Only