missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ SP ImpRes BioProcess Media

Catalog No. 45002563
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002563 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002563 Supplier Cytiva Supplier No. 17546810

missing translation for 'validMainframeMsg'

A strong cation exchange BioProcess medium for efficient and cost-effective protein purification.

  • Chemically stable with commonly used aqueous buffers
  • High manufacturing productivity
  • Sulfate group ligand
  • High-flow agarose matrix
  • Flexible process design due to large operational window of flow rates and bed heights

Avoid using oxidizing agents and cationic detergents

Developed and supported for large-scale chromatography; used for high-throughput intermediate purification and polishing steps of a wide range of biomolecules

Certifications

Certificate of Analysis

Specifications

Quantity 25 mL
Stability 1 M NaOH, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
For Use With (Application) For Large Scale PurifiCatIon of Proteins
Storage Requirements 4°C to 30°C
Binding Capacity >70 mg Lysozyme, >95 mg BSA
Product Type Capto SP ImpRes Media
Flow Rate 220 cm/Hoμr
Particle Size 36 to 44 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Ionic Capacity 0.13 to 0.16 mM Hl+/mL
Includes Certificate of Analysis
Ion Exchange Type Strong CatIon Exchanger
Max. Pressure 300 kPa
Matrix High-flow Agarose
pH Range 3 to 14
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Sulphonate Group
Show More Show Less