missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ Multimodal (MMC) Chromatography Media

Catalog No. 45002952
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002952 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002952 Supplier Cytiva Supplier No. 17531702

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to solve specific purification challenges at both high and low ionic strengths

  • Composed of rigid, high-flow agarose matrix and novel multimodal ligand
  • Used to bind proteins at conductivity of feed material
  • Ligand contains carboxylic acid group and exhibit the properties of weak cation exchanger
  • High dynamic binding capacity at high conductivity
  • High-volume throughput cuts process times
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Dynamic binding capacity at 10% breakthrough as measured at a residence time of 2 min, 300cm/h in a Tricorn(TM) 5/100 column with 10cm bed height in 50mM Sodium acetate, pH 4.75, 250mM NaCl.

Warranty

Store at 4° to 30°C in 20% Ethanol

Specifications

Quantity 100 mL
For Use With (Application) Used to Solve Specific PurifiCatIon Challenges at Both High and Low Ionic Strengths
Stability 1 M Acetic Acid, 1 M Sodium Hydroxide, 8 M Urea, 6 M Guanidine Hydrochloride, and 70% Ethanol
Storage Requirements 4°C to 30°C
Binding Capacity >45 mg BSA/mL medium (at 30 mS/cm2)
Product Type Capto MMC Media
Flow Rate 600 cm/Hoμr
Particle Size 75 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Ionic Capacity 0.07 to 0.09 mM H+/mL
Ion Exchange Type Multimodal Weak CatIon Exchanger
Max. Pressure 300 kPa
Matrix Highly cross-linked agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type Carboxylic Acid Group
Show More Show Less