missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ MMC ImpRes Multimodal BioProcess™ Chromatography Medium

Catalog No. 45002507
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002507 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002507 Supplier Cytiva Supplier No. 17371602

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules

  • High yields achieved through high-resolution beads and ligand selectivity
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • Enables use of platform approach to MAb process development
  • Displays broad pH/salt operational window

Avoid using oxidizing agents, cationic detergents.

Specifications

Quantity 100 mL
Stability 1 M Acetic Acid, 1 M NaOH, 2 M NaCl, 5% 1-Propanol, 30% Isopropanol, 70% Ethanol
For Use With (Application) For High Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules
Storage Requirements 4°C to 30°C
Anion Exchange Capacity 25 to 39 μM/mL
Binding Capacity 60 to 90 mg Mab/mL
Product Type Capto MMC ImpRes Media
Particle Size 40 μm
Max. Pressure 300 kPa
Matrix High-flow Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Multimodal Weak CatIon Exchanger
Format Bulk Media
Show More Show Less