missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ Blue Chromatography Media

Catalog No. 45002994
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002994 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002994 Supplier Cytiva Supplier No. 17544801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An affinity medium for capture of proteins and enzyme purification on large and small scale

This product is chemically stable for cleaning with strong agarose base matrix.

  • Chemical stability ensures tolerance of harsh solvents used in repeated CIP and sanitization procedures
  • Rigid agarose base matrix allows high flow rates and processing of large sample volumes with no reduction in binding capacity
  • Can be autoclaved repeatedly
  • Average particle size of 75μm

For purification of enzymes, capture of albumin and recombinant proteins at laboratory and process scales

Specifications

Quantity 25 mL
Stability Stable in commonly used aqueous Buffers: 0.5 M NaOH, 8 M Urea, 6 M Guanidine Hydrochloride
For Use With (Application) For Enzyme PurifiCatIon, Capture of Albumin
Storage Requirements 2°C to 8°C
Binding Capacity >24 mg HSA/mL mediμm
Product Type Capto Blue Media
Flow Rate 600 cm/Hoμr
Particle Size 75 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure 300 kPa
Matrix Highly Cross-Linked Agarose
pH Range 2 to 13
Storage Buffer 20% Ethanol
Ligand Type Cibacron Blue
Show More Show Less