missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ adhere ImpRes Multimodal BioProcess™ Chromatography Medium

Catalog No. 45002505
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002505 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002505 Supplier Cytiva Supplier No. 17371502

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Certificate of Analysis

For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules

The strong anion exchange multimodal ligand displays high selectivity compared with traditional ion exchanger, which allows the possibility to solve challenging purification problems. Main contaminants in MAb processes such as DNA, host cell proteins (HCP), leached protein A, aggregates, and viruses are efficiently separated.

  • High-resolution beads and ligand selectivity ensures high yield
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • Enables use of platform approach to MAb process development
  • Allows separation of MAb charged variants

Avoid using oxidizing agents, anionic detergents.

Specifications

Quantity 100 mL
For Use With (Application) For High Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules
Stability 1 M Acetic Acid, 1 M NaOH, 2 M NaCl, 5% 1-Propanol, 30% Isopropanol, 70% Ethanol
Storage Requirements 4°C to 30°C
Anion Exchange Capacity 0.08 to 0.11 mM Cl-/mL
Binding Capacity 45 to 85 mg Mab/mL
Product Type Capto Adhere ImpRes Media
Particle Size 40 μm
Includes Certificate of Analysis
Max. Pressure 300 kPa
Matrix High-flow Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Multimodal Strong AnIon Exchanger
Show More Show Less