missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPSO sodium salt, 98%

Catalog No. aab2520222
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2520222 100 g
AAB2520214 25 g
AAB2520236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2520222 Supplier Thermo Scientific Chemicals Supplier No. B2520222
Only null left

CAS: 102601-34-3 | C9H12NO4S | 230.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102601-34-3
Molecular Formula C9H12NO4S
Molecular Weight (g/mol) 230.26
MDL Number MFCD00070063
InChI Key CLEVULOKVPBHCU-VIFPVBQESA-M
Synonym capso sodium salt, sodium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, 3-cyclohexylamino-2-hydroxypropanesulfonic acid sodium salt, capso sodium, capso, 0.2m buffer solution, ph 8.5, capso, 0.2m buffer solution, ph 9.0, capso, 0.2m buffer solution, ph 9.5, sodium 3-cyclohexylamino-2-hydroxypropanesulfonate, sodium 3-cyclohexylamino-2-hydroxy-propane-1-sulfonate, potassium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate
PubChem CID 25000353
IUPAC Name sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
SMILES O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O

Specifications

Quantity 100 g
Sensitivity Hygroscopic
Formula Weight 259.3
Percent Purity 98%
Chemical Name or Material CAPSO sodium salt
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only