accessibility menu, dialog, popup

UserName

CAS RN 102601-34-3

IUPAC Name: (2S)-2-hydroxy-3-(phenylamino)propane-1-sulfonate
Synonyms: (2S)-2-hydroxy-3-(phenylamino)propane-1-sulfonate
Molecular Weight (g/mol): 230.26
Molecular Formula: C9H12NO4S
InChi Key: CLEVULOKVPBHCU-VIFPVBQESA-M
SMILES: O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)