missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capsanthin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 465-42-9 |
| Linear Formula | C40H56O3 |
| Synonym | (3 R,3′S,5′R)-3,3′-Dihydroxy-beta,kappa-caroten-6′-one |
| Type | Analytical Standard |
| Quantity | 1 mg |
| Formula Weight | 584.87 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Analytical Standard |