missing translation for 'onlineSavingsMsg'
Learn More

Capsaicin, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187408
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019937 10 mg
111018501 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019937 Supplier Merck Emd Millipore Supplier No. 1208410MGF
Only null left

Superior quality of pure E-Capsaicin, with a guaranteed assay of =99.0% by HPLC.

Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

Description Analytical standard for food analysis
CAS 404-86-4
Flash Point 113°C
Physical Form Neat
Linear Formula (CH3)2 CHCH=CH(CH2)4 CONHCH2C6H3-4-(OH)-3-(OCH3)
MDL Number MFCD00017259
Melting Point 62°C to 65°C
Synonym 8-Methyl-N-vanillyl-trans-6-nonenamide
Solubility Insoluble in water
Type Analytical Standard
Quantity 10 mg
Formula Weight 305.41
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Show More Show Less