missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa584411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584411 Supplier Invitrogen™ Supplier No. PA584411
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: SEEESDEIVE EESGKEVLGS APSGARLSPS RTSEGS Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 89%.

This gene encodes a novel neural/endocrine-specific cytosolic and peripheral membrane protein required for the Ca2+-regulated exocytosis of secretory vesicles. The protein acts at a stage in exocytosis that follows ATP-dependent priming, which involves the essential synthesis of phosphatidylinositol 4,5-bisphosphate P2). Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described.

Specifications

Antigen CAPS1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CADPS
Gene Accession No. Q9ULU8
Gene Alias AU067781; Ca++-dependent secretion activator; Ca>2+>dependent activator protein for secretion; Ca>2+-dependent activator protein for secretion; Ca2+ dependent secretion activator; Ca2+-dependent activator protein; Ca2+-dependent activator protein for secretion; Ca2+-dependent secretion activator; Ca2+-regulated cytoskeletal protein; Cadps; CADPS1; calcium dependent secretion activator; calcium-dependent activator protein for secretion 1; Calcium-dependent secretion activator 1; Caps; Caps1; CAPS-1; Caps2; KIAA1121; mKIAA1121; rCAPS; UNC-31
Gene Symbols CADPS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CADPS. Recombinant protein control fragment (Product #RP-103351).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less