Learn More
Fisherbrand™ Caps with Pierceable Cross Cut for Auto-Analyzer Sample Cups
Catalog No. 22170181
Change view
Click to view available options
Type: Criss-Cross Cut Cap
Catalog No.
22170181 Type: Criss-Cross Cut Cap
1 options
Catalog No. 22170181 Supplier Fisherbrand™ Supplier No. 113133PLFH
Only null left
Produced from polyethylene (PE), these caps are designed specifically for use on our popular line of multi-purpose sample cups.
  • The caps are available with a solid or cross-cut pierceable top.
  • All caps snap securely in place on each of the sample cups.
Specifications
Product Type Pierceable Stopper
Material Polyethylene
Quantity 1000/Cs.
Sterility Non-sterile
For Use With (Equipment) Fisherbrand Auto-Analyzer Cups
Disposable Single-use