missing translation for 'onlineSavingsMsg'
Learn More

Caprylic acid, >99.5%, MP Biomedicals™

Catalog No. p-4403710
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10039190 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10039190 Supplier MP Biomedicals Inc Supplier No. 0210039190
Only null left

Caprylic acid is the common name for the eight-carbon saturated fatty acid known by the systematic name octanoic acid. It is found naturally in the milk of various mammals, and it is a minor constituent of coconut oil and palm kernel oil

Chemical Identifiers

CAS 124-07-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00004429
InChI Key WWZKQHOCKIZLMA-UHFFFAOYSA-N
Synonym caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
PubChem CID 379
ChEBI CHEBI:28837
IUPAC Name octanoic acid
SMILES CCCCCCCC(=O)O

Specifications

Color Colorless
Density 0.91g/cm3 at 20°C
Assay Percent Range >99.5%
Quantity 500 mL
Formula Weight 144.2
Physical Form Liquid
Chemical Name or Material Caprylic acid
EINECSNumber 204-677-5
Recommended Storage Store at room temperature (15° to 30°C).