missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Burdick & Jackson™ Capping Reagent, 84/16 THF/NMI (v/v)

Catalog No. 60048384
Change view
Click to view available options
Quantity:
4 x 2 L
Packaging Type:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
60048384 4 x 2 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048384 Supplier Honeywell Burdick & Jackson™ Supplier No. BR6502
Only null left

BioSyn™, for DNA/RNA synthesis

Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis

  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Specifications

CAS 616-47-7
Content And Storage Room Temperature
Density 0.910 g/cm3
Packaging Type Glass Bottle
Quantity 4 x 2 L
Form Liquid