missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Burdick & Jackson™ Capping Reagent, 80/20 Acetonitrile/NMI (v/v)
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
Specifications
| CAS | 75-05-8 |
| Content And Storage | Room Temperature |
| Density | 0.827 g/cm3 |
| Packaging Type | GLASS BOTTLE, GL45 |
| Quantity | 4 x 2.5 L |
| Form | Liquid |