missing translation for 'onlineSavingsMsg'
Learn More

Solstice Capping Reagent, 80/20 Acetonitrile/NMI (v/v)

Catalog No. 60048389
Change view
Click to view available options
Quantity:
2 x 1 L
4 x 2.5 L
Packaging Type:
Glass Bottle
GLASS BOTTLE, GL45
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
60048389 2 x 1 L Glass Bottle
60048390 4 x 2.5 L GLASS BOTTLE, GL45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60048389 Supplier Solstice Supplier No. BR6541

BioSyn™, for DNA/RNA synthesis

Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis

  • Solstice BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Specifications

CAS 75-05-8
Content And Storage Room Temperature
Density 0.827 g/cm3
Packaging Type Glass Bottle
Quantity 2 x 1 L
Form Liquid