missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Capitol Vial Universal Test Tube Plug Caps

Description
Innovative flanged-thread design allows cap to descend into the tube until a friction fit is secure and plug is stationary
- Fit tubes with 12 to 16 mm diameter
- Hollow top finish with ribs for easy retrieval
Specifications
Specifications
| Product Type | Test Tube Plug Cap |
| Material | LDPE |
| Color | Yellow |
| Quantity | 10,000/Cs. |
| Sterility | Non-sterile |
| For Use With (Equipment) | Fits Tubes with Inner Diameters of 12 to 16 mm |