Learn More
Thermo Scientific™ Capit-All Flex Cap Driver Cassettes
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format. It is recommended to be replaced after 10,000 decapping cycles.
$19,468.91
| Catalog Number | Mfr. No. | Compatible Storage Tubes | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Compatible Storage Tubes | Price | Quantity & Availability | |||||
|
14377140
|
Thermo Scientific™
4130MATNUN |
Matrix and Nunc tubes in 96 or 48 formats |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format. It is recommended to be replaced after 10,000 decapping cycles.
- Cap driver cassette compatible with Thermo Scientific Capit-All Flex Automated Decapper (P/N 4120-FLEX)
- Tube compatibility: Matrix and Nunc cryotubes in 96 and 48 formats; Azenta FluidX tubes with internal threads in 96 format
- Replace after 10,000 cycles
Specifications
| 96-Channel | |
| Decapping Head |