Learn More
Thermo Scientific™ Capit-All Flex Cap Driver Cassettes
Catalog No. 14372465
Change view
Click to view available options
Compatible Storage Tubes:
Azenta 96 Format External Thread
Azenta 96 Format Internal Thread
Matrix and Nunc tubes in 96 or 48 formats
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Compatible Storage Tubes
14372465 Azenta 96 Format External Thread
14372466 Azenta 96 Format Internal Thread
14377140 Matrix and Nunc tubes in 96 or 48 formats
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14372465 Supplier Thermo Scientific™ Supplier No. 4130FDXE
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format. It is recommended to be replaced after 10,000 decapping cycles.
  • Cap driver cassette compatible with Thermo Scientific Capit-All Flex Automated Decapper (P/N 4120-FLEX)
  • Tube compatibility: Matrix and Nunc cryotubes in 96 and 48 formats; Azenta FluidX tubes with internal threads in 96 format
  • Replace after 10,000 cycles
Specifications
Compatible Storage Tubes Azenta 96 Format External Thread
Decapping Format 96-Channel
Type Decapping Head