missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific Capinsert™ Cap Inserts
Description
- Simplifies sample identification
- Press into place
- Frosted writing area on top provides space for additional identifying markings
- Made of polypropylene
Order Info
Available in 11 colors. Assorted packages contain 100 of each color.
Specifications
Specifications
| Pre-sterilized | No |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Color | Green |
| For Use With (Application) | Used to color code and write on frosted area for sample identification |
| No. per Pack | 500 Pk. |
| Sterility | Non-sterile |
| Tube Material | Polypropylene |
| For Use With (Equipment) | Micrewtubes Microcentrifuge Tubes and Other Products |