missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Capinsert™ Cap Inserts

Catalog No. 056702
Change view
Click to view available options
Color:
Gray
Green
Orange
Pink
Red
Violet
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
056702 Red
22045361 Green
22045363 Orange
22045360 Gray
22045421 Pink
22045423 Violet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 056702
Only null left

Capinsert™ Cap Inserts are used to color code a multitude of products such as tubes and vials. They are inserted on top of the closure and has a write-on frosted area for sample identification.

  • Simplifies sample identification
  • Press into place
  • Frosted writing area on top provides space for additional identifying markings
  • Made of polypropylene

Order Info

Available in 11 colors. Assorted packages contain 100 of each color.

Specifications

Pre-sterilized No
Autoclavable Autoclavable
Material Polypropylene
Color Red
For Use With (Application) Used to color code and write on frosted area for sample identification
No. per Pack 500 Pk.
Sterility Non-sterile
Tube Material Polypropylene
For Use With (Equipment) Micrewtubes Microcentrifuge Tubes and Other Products