missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Capillary Cutter, 310 Genetic Analyzer

Catalog No. 401958
Encompass
Change view
Click to view available options
Quantity:
2 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401958 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401958 Supplier Applied Biosystems™ Supplier No. 401958

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This is a capillary cutter for use with 310 genetic analyzer capillaries. Shortened capillaries can be used for short run modules.

This is a capillary cutter for use with 310 Genetic Analyzer capillaries. Shortened capillaries can be used for short run modules.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Product Type Capillary Cutter
Quantity 2 Each
Shipping Condition Room Temperature

For research use only