missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAPG Monoclonal Antibody (K52030_15B12), Invitrogen™

Catalog No. MA555907
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555907 50 μL
MA556287 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555907 Supplier Invitrogen™ Supplier No. MA555907
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: RDLALAIRDS ERQGKAQVEI VTDGEEPAEM IQVLGPKPAL KEGNPEEDLT ADKANAQAAA LYKVSDATGQ MNLTKVADSS

This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in non-muscle cells. Alternatively spliced transcript variants have been observed, but have not been fully described.

Specifications

Antigen CAPG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K52030_15B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CAPG
Gene Accession No. P40121
Gene Alias Actin regulatory protein CAP-G; actin-capping protein GCAP39; actin-regulatory protein CAP-G; AFCP; Capg; capping actin protein, gelsolin like; capping protein (actin filament), gelsolin-like; epididymis secretory protein Li 66; gCap39; gelsolin-like capping protein; HEL-S-66; macrophage capping protein; macrophage-capping protein; mbh1; MCP; myc basic motif homolog 1
Gene Symbols CAPG
Host Species Mouse
Immunogen Recombinant human CAPG (immunogen range is 191-270).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less