missing translation for 'onlineSavingsMsg'
Learn More

Capecitabine 98.0+%, TCI America™

Catalog No. p-7125699
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
C28781G 1 g
C28785G 5 g
2 options

Catalog No. C28781G

Supplier: TCI America C28781G

Only null left

Chemical Identifiers

CAS 154361-50-9
Molecular Formula C15H22FN3O6
Molecular Weight (g/mol) 359.354
MDL Number MFCD00930626
InChI Key GAGWJHPBXLXJQN-UORFTKCHSA-N
Synonym capecitabine, xeloda, capiibine, capecitibine, caxeta, xabine, capecitabine usan, capecitabina, capecitabinum, capecitabin
PubChem CID 60953
ChEBI CHEBI:31348
IUPAC Name pentyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
SMILES CCCCCOC(=O)NC1=NC(=O)N(C=C1F)C2C(C(C(O2)C)O)O

Specifications

Melting Point 123°C
Color White
Quantity 1 g
Formula Weight 359.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Capecitabine
RTECSNumber HA3852500
TSCA No
Recommended Storage Refrigerator