missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAP1 Recombinant Rabbit Monoclonal Antibody (23GB3020)
GREENER_CHOICE

Catalog No. MA553318
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553318 100 μL
MA553319 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553318 Supplier Invitrogen™ Supplier No. MA553318
Only null left

Rabbit Recombinant Monoclonal Antibody

CAP1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Adenylyl cyclase-associated protein 1 is an enzyme that in humans is encoded by the CAP1 gene. This gene is mapped to 1p34. 2. The protein encoded by this gene is related to the S. cerevisiae CAP protein, which is involved in the cyclic AMP pathway. And the human protein is able to interact with other molecules of the same protein, as well as with CAP2 and actin. Alternatively spliced transcript variants have been identified.

Specifications

Antigen CAP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3020
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CAP1
Gene Accession No. Q01518
Gene Alias adenylate cyclase associated protein 1; adenylyl cyclase-associated CAP protein, yeast homolog 1; adenylyl cyclase-associated protein 1; CAP; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP, adenylate cyclase-associated protein 1 (yeast); CAP1; CAP1-PEN; cyclase-associated protein homologue; Mch1; RP11-115D7.1; testis secretory sperm-binding protein Li 218p
Gene Symbols CAP1
Host Species Rabbit
Immunogen A synthesized peptide derived from CAP1 (12-45AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10487
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less