missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canthaxanthin (trans), ≥95.0%, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 514-78-3 |
| Physical Form | Neat |
| Linear Formula | C40H52O2 |
| MDL Number | MFCD00016364 |
| Melting Point | 201.6°C to 201.7°C |
| Synonym | beta-Carotin-4,4 ′-dione; E 161 g |
| Type | Analytical Standard |
| Quantity | 2 mg |
| Formula Weight | 564.84 |
| Percent Purity | ≥95.0% |
| Show More |