missing translation for 'onlineSavingsMsg'
Learn More

Cantharidin 98.0+%, TCI America™

Catalog No. p-7124178
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C001925MG 25 mg
C0019100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C001925MG Supplier TCI America Supplier No. C001925MG
Only null left

Chemical Identifiers

CAS 56-25-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00134968
InChI Key DHZBEENLJMYSHQ-XCVPVQRUSA-N
Synonym cantharidin, cantharone, cantharidine, kantaridin, kantharidin, cantharides camphor, kantharidin german, caswell no. 157, 1,2-dimethyl-3,6-epoxyperhydrophthalic anhydride, unii-igl471wq8p
PubChem CID 5944
ChEBI CHEBI:64213
SMILES CC12C3CCC(C1(C(=O)OC2=O)C)O3

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 25 mg
UN Number 2811
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cantharidin
RTECSNumber RN8575000
TSCA Yes