accessibility menu, dialog, popup

UserName

CAS RN 56-25-7

IUPAC Name: (1R,2S,6R,7S)-2,6-dimethyl-4,10-dioxatricyclo[5.2.1.0²,⁶]decane-3,5-dione
Synonyms: cantharidin kantharidin cantharone cantharidine 斑蝥素
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: DHZBEENLJMYSHQ-XCVPVQRUSA-N
SMILES: C[C@]12[C@@H]3CC[C@@H](O3)[C@@]1(C)C(=O)OC2=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)