missing translation for 'onlineSavingsMsg'
Learn More

Cantharidin 98.0+%, TCI America™

Catalog No. c001925mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
25 mg
Catalog No. Quantity
C001925MG 25 mg
C0019100MG 100 mg
2 options

Catalog No. C001925MG

Supplier: TCI America C001925MG

Only null left

Chemical Identifiers

CAS 56-25-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00134968
InChI Key DHZBEENLJMYSHQ-XCVPVQRUSA-N
Synonym cantharidin, cantharone, cantharidine, kantaridin, kantharidin, cantharides camphor, kantharidin german, caswell no. 157, 1,2-dimethyl-3,6-epoxyperhydrophthalic anhydride, unii-igl471wq8p
PubChem CID 5944
ChEBI CHEBI:64213
SMILES CC12C3CCC(C1(C(=O)OC2=O)C)O3

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 25 mg
UN Number 2811
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cantharidin
RTECSNumber RN8575000
TSCA Yes