missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Canine RAGE ELISA Kit
ELISA
Supplier: Invitrogen™ EC14RB
Description
Canine RAGE quantitates canine RAGE in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant canine RAGE.
The Receptor for Advanced Glycation End-products (RAGE) is a gene located on human chromosome 6p21.3, encoding a transmembrane receptor belonging to the immunoglobulin superfamily. RAGE is expressed in various tissues, with significant levels in the lungs, and plays a crucial role in cellular signaling and inflammation. As a receptor, RAGE binds multiple ligands, including advanced glycation end-products (AGEs), amyloid-beta peptide, high mobility group box 1 (HMGB1), and S100/calgranulin proteins, facilitating diverse pathological processes like inflammation, cancer progression, and neurodegeneration. The interaction between RAGE and its ligands triggers intracellular signaling pathways such as NF-kB activation, leading to inflammatory responses and oxidative stress. In the context of chronic diseases like diabetes, Alzheimer's, and cardiovascular diseases, RAGE is a critical mediator, linking metabolic disturbance to cellular dysfunction. Therapeutic targeting of RAGE signaling is under investigation, aiming to mitigate its contribution to inflammatory and degenerative diseases.Chemical Identifiers
| advanced glycation end product receptor; advanced glycation end-products receptor; advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; AGER; Rage; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE/AGER; RAGE-4 ORF3; receptor for advanced glycation end-products variant 20; receptor for advanced glycosylation end products; SCARJ1; sRAGE | |
| 16 pg/mL | |
| ELISA | |
| Absorbance Microplate Reader | |
| AGER | |
| <10% | |
| Biotin | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| RAGE | |
| 4 hr 45 min |
| 16-4000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 403168 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
| 1 hr 20 min |
Specifications
| advanced glycation end product receptor; advanced glycation end-products receptor; advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; AGER; Rage; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE/AGER; RAGE-4 ORF3; receptor for advanced glycation end-products variant 20; receptor for advanced glycosylation end products; SCARJ1; sRAGE | |
| 16 pg/mL | |
| Sandwich ELISA Kit | |
| Plasma, Serum, Supernatant | |
| Absorbance Microplate Reader | |
| 403168 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
| 1 hr 20 min |
| 16-4000 pg/mL | |
| HRP | |
| ELISA | |
| Canine | |
| advanced glycation end product receptor; advanced glycation end-product receptor; advanced glycation end-products receptor; advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; advanced glycosylation end-product specific receptor; Ager; MAPK/MAK/MRK overlapping kinase; MOK; MOK protein kinase; RAGE; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE/AGER; RAGE1; RAGE-1; RAGE-4 ORF3; receptor for advanced glycation endproducts; receptor for advanced glycation end-products variant 20; receptor for advanced glycosylation end products; receptor of advanced glycosylation end products of proteins; immunoglobulin superfamily; MHC class II; MHC class III; renal cell carcinoma antigen; renal tumor antigen 1; SCARJ1; sRAGE; STK30advanced glycation end product receptor; advanced glycation end-product receptor; advanced glycation end-products receptor; advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; advanced glycosylation end-product specific receptor; Ager; MAPK/MAK/MRK overlapping kinase; MOK; MOK protein kinase; RAGE; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE/AGER; RAGE1; RAGE-1; RAGE-4 ORF3; receptor for advanced glycation endproducts; receptor for advanced glycation end-products variant 20; receptor for advanced glycosylation end products; receptor of advanced glycosylation end products of proteins; immunoglobulin superfamily; MHC class II; MHC class III; renal cell carcinoma antigen; renal tumor antigen 1; SCARJ1; sRAGE; STK30; | |
| AGER | |
| <10% | |
| Biotin | |
| RUO | |
| Wet or Dry Ice | |
| RAGE | |
| 4 hr 45 min |