missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Canine IGFBP-2 ELISA Kit
ELISA
Supplier: Invitrogen™ EC9RB
Description
Canine IGFBP-2 quantitates canine IGFBP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant canine IGFBP-2.
IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.Specifications
| AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; Igfbp-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36 kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753 | |
| 0.055 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| IGFBP2 | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.055 to 12.50 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 488516 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| IGFBP2 | |
| 4 hrs 45 mins |