missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMSAP1L1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302161AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
CAMSAP2 gene ontology annotations related to this gene include neuron projection development.Specifications
| CAMSAP1L1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q08AD1 | |
| CAMSAP2 | |
| Between 375 and 425 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CAMSAP2 | |
| calmodulin regulated spectrin-associated protein 1-like 1, calmodulin regulated spectrin-associated protein family, member 2, calmodulin-regulated spectrin-associated protein 1-like protein 1, CAMSAP1L1, KIAA1078 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23271 | |
| 4° C | |
| Liquid |