missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Camphorquinone 98.0+%, TCI America™

$112.02 - $318.97

Chemical Identifiers

CAS 10373-78-1
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00064160
InChI Key VNQXSTWCDUXYEZ-UHFFFAOYSA-N
Synonym camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
PubChem CID 25208
ChEBI CHEBI:34607
IUPAC Name 4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
SMILES CC1(C2CCC1(C(=O)C2=O)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
C00145G
View Documents Promotion Details
N/A 5 g
Each for $112.02
Only null left
 
C001425G
View Documents Promotion Details
N/A 25 g
Each for $318.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10373-78-1
166.22
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
25208
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
C10H14O2
MFCD00064160
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
CHEBI:34607
CC1(C2CCC1(C(=O)C2=O)C)C

Specifications

10373-78-1
Yellow
MFCD00064160
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
25208
166.22
Crystalline Powder
203°C
C10H14O2
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
CC1(C2CCC1(C(=O)C2=O)C)C
166.22
CHEBI:34607
≥98.0% (GC)
(+/-)-Camphorquinone
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents