missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Camphorquinone 98.0+%, TCI America™

Supplier:  TCI America C001425G

Quantity 25 g

 View more versions of this product

Catalog No. C001425G


Only null left

Description

Description

Specifications

Chemical Identifiers

10373-78-1
166.22
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
25208
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
C10H14O2
MFCD00064160
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
CHEBI:34607
CC1(C2CCC1(C(=O)C2=O)C)C

Specifications

(+/-)-Camphorquinone
203°C
C10H14O2
MFCD00064160
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
25208
166.22
Crystalline Powder
10373-78-1
Yellow
25 g
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
CC1(C2CCC1(C(=O)C2=O)C)C
166.22
CHEBI:34607
≥98.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions