missing translation for 'onlineSavingsMsg'
Learn More

(+)-Camphoric Acid 98.0+%, TCI America™

Catalog No. c0012100g
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0012100G 100 g
C001225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0012100G Supplier TCI America Supplier No. C0012100G
Only null left

Chemical Identifiers

CAS 124-83-4
Molecular Formula C10H14O4-2
Molecular Weight (g/mol) 198.218
MDL Number MFCD00001375
InChI Key LSPHULWDVZXLIL-LDWIPMOCSA-L
Synonym d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
PubChem CID 6918944
IUPAC Name (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
SMILES CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C

Specifications

Melting Point 187°C
Color White
Formula Weight 200.23
Percent Purity ≥98.0% (GC,T)
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material (+)-Camphoric Acid
EINECSNumber (4)-0308
TSCA Yes