accessibility menu, dialog, popup

UserName

CH3H3C(S)(R)H3COOOO

CAS RN 124-83-4

IUPAC Name: (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
Synonyms: (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: LSPHULWDVZXLIL-LDWIPMOCSA-L
SMILES: CC1(C)[C@H](CC[C@@]1(C)C([O-])=O)C([O-])=O
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)