missing translation for 'onlineSavingsMsg'
Learn More

(+)-Camphor 98.0+%, TCI America™

Catalog No. p-7124171
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C001025G 25 g
C0010500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C001025G Supplier TCI America Supplier No. C001025G
Only null left

Chemical Identifiers

CAS 464-49-3
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149
InChI Key DSSYKIVIOFKYAU-XVKPBYJWSA-N
Synonym +-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone
PubChem CID 159055
ChEBI CHEBI:15396
IUPAC Name (1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
SMILES CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2

Specifications

Melting Point 178°C
Color White
Boiling Point 204°C
Quantity 25 g
UN Number 2717
Formula Weight 152.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Camphor
EINECSNumber (4)-0308&(4)-0601
RTECSNumber EX1260000
TSCA Yes