missing translation for 'onlineSavingsMsg'
Learn More

(±)-Camphor, 96%

CAS: 76-22-2 | C10H16O | 152.24 g/mol

Supplier:  Thermo Scientific Chemicals A109360E

Quantity 2500 g

 View more versions of this product

Catalog No. AAA109360E


Only null left

Description

Description

Camphor is used in the preparation of mothballs. It acts as a plasticizer for nitrocellulose, and an ingredient for fireworks and explosive munitions. It is useful in the treatment of sprains, swellings and inflammation. It is also used to synthesize carbon nanotubes by chemical vapor deposition process of camphor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphor is used in the preparation of mothballs. It acts as a plasticizer for nitrocellulose, and an ingredient for fireworks and explosive munitions. It is useful in the treatment of sprains, swellings and inflammation. It is also used to synthesize carbon nanotubes by chemical vapor deposition process of camphor.

Solubility
Soluble in acetone, ethanol, diethylether, chloroform and acetic acid.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and chlorinated solvents.
Specifications

Chemical Identifiers

76-22-2
152.24
DSSYKIVIOFKYAU-UHFFFAOYNA-N
2537
4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
C10H16O
MFCD00074738,MFCD00064149
camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon
CHEBI:36773
CC1(C)C2CCC1(C)C(=O)C2

Specifications

(±)-Camphor
172°C to 176°C
204°C
Camphor-like
C10H16O
MFCD00074738,MFCD00064149
1907611
camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon
DSSYKIVIOFKYAU-UHFFFAOYNA-N
4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
2537
152.24
76-22-2
0.992
64°C (147°F)
96%
2500 g
UN2717
14,1732
Soluble in acetone,ethanol,diethylether,chloroform and acetic acid.
CC1(C)C2CCC1(C)C(=O)C2
152.24
CHEBI:36773
96%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR

EINECSNumber : 200-945-0

RTECSNumber : EX1225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions